AMG-1 98%

Supplier: Ambeed
Warning

A1212551-10MG A1212551-1MG A1212551-50MG A1212551-5MG
76678-078EA 110 USD
76678-078 76678-080 76678-082 77620-896
AMG-1 98%
AMG-1

Caution: for Research Use Only

Formula: C₃₁H₂₉FN₄O₅
MDL Number: MFCD22631502
CAS Number: 913376-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR