Chloro(1,5-cyclooctadiene)iridium(I) dimer 98% (57.2% Ir)
Supplier: Ambeed
|
Warning
|
Synonyms:
[Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer
A1194534-1G
A1194534-25G
76677-266EA
225
USD
76677-266
76677-268
77866-509
Chloro(1,5-cyclooctadiene)iridium(I) dimer 98% (57.2% Ir)
Chloro(1,5-cyclooctadiene)iridium (I) dimer
Caution: for Research Use Only
Formula:
C₁₆H₂₄Cl₂Ir₂ MW: 671.70 g/mol |
MDL Number:
MFCD00012414 CAS Number: 12112-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...