Bisindoylmaleimide X hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Bisindolylmaleimide X HCl, Ro 31-8425, 3-(8-(Aminomethyl)-6,7,8,9-tetrahydropyrido[1,2-a]indol-10-yl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione hydrochloride

A1159144-10MG A1159144-25MG A1159144-5MG
76675-470EA 223 USD
76675-470 76675-472 76675-474
Bisindoylmaleimide X hydrochloride 98%
Bisindoylmaleimide X hydrochloride

Caution: for Research Use Only

Formula: C₂₆H₂₄N₄O₂·HCl
MDL Number: MFCD04972010
CAS Number: 145317-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR