Manganese(II) sulfide 99.9%

Supplier: Ambeed
Danger

A1150350-25G A1150350-500G A1150350-1000G
76674-262EA 6 USD
76674-262 77620-116 77620-114
Manganese(II) sulfide 99.9%
Manganese (II) sulphide

Caution: for Research Use Only

Formula: MnS
MW: 87.00 g/mol
MDL Number: MFCD00014210
CAS Number: 18820-29-6
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR