Cladribine 98%

Supplier: Ambeed
Danger

Synonyms: (2R,3S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol, CdA, 2-Chloro-2'-deoxyadenosine, NSC105014, RWJ 26251

A410468-10MG A410468-1G A410468-1MG A410468-250MG A410468-50MG A410468-100MG A410468-5G
76669-790EA 28.65 USD
76669-790 76669-792 76669-794 76669-796 76669-798 77598-468 77598-470
Cladribine 98%
Cladribine

Caution: for Research Use Only

Formula: C₁₀H₁₂ClN₅O₃
MW: 285.7 g/mol
MDL Number: MFCD00153939
CAS Number: 4291-63-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR