3-Amino-3-phenyl-1-propanol 98+%

Supplier: Ambeed
Danger

A1473847-100G A1473847-25G A1473847-5G
76663-174EA 130 USD
76663-174 76663-176 76663-178
3-Amino-3-phenyl-1-propanol 98+%
3-Amino-3-phenyl-1-propanol

Caution: for Research Use Only

Formula: C₉H₁₃NO
MDL Number: MFCD00801124
CAS Number: 14593-04-5
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR