(11bS)-N,N-Bis[(1S)-1-(4-methoxyphenyl)ethyl]-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine 95%

Supplier: Ambeed
Warning

A1370380-100MG A1370380-1G A1370380-250MG
76659-564EA 119 USD
76659-564 76659-566 76659-568
(11bS)-N,N-Bis[(1S)-1-(4-methoxyphenyl)ethyl]-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine 95%
(11bS)-N,N-Bis[(1S)-1-(4-methoxyphenyl)ethyl]-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine

Caution: for Research Use Only

Formula: C₃₈H₃₄NO₄P
CAS Number: 850796-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR