(R)-1-(9H-Fluoren-9-yl)ethanol 97%

Supplier: Ambeed
Warning

A1370332-100MG A1370332-250MG
76659-560EA 70 USD
76659-560 76659-562
(R)-1-(9H-Fluoren-9-yl)ethanol 97%
(R)-1-(9H-Fluoren-9-yl)ethanol

Caution: for Research Use Only

Formula: C₁₅H₁₄O
CAS Number: 107474-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR