Cyanine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Isopentyl-4-((1-isopentylquinolin-4(1H)-ylidene)methyl)quinolin-1-ium iodide, Quinoline Blue

A987977-250MG A987977-1G A987977-100MG
76657-154EA 46.68 USD
76657-154 77599-380 77618-708
Cyanine 98%
Cyanine

Caution: for Research Use Only

Formula: C₂₉H₃₅IN₂
MW: 538.52 g/mol
MDL Number: MFCD00050330
CAS Number: 523-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR