9,10-Bis[N-(2-naphthyl)anilino]anthracene 98% (by HPLC, total nitrogen)

Supplier: Ambeed
Warning

A940316-100MG A940316-1G A940316-250MG
76655-770EA 32 USD
76655-770 76655-772 76655-774
9,10-Bis[N-(2-naphthyl)anilino]anthracene 98% (by HPLC, total nitrogen)
9,10-Bis[N-(2-naphthyl)anilino]anthracene

Caution: for Research Use Only

Formula: C₄₆H₃₂N₂
MDL Number: MFCD12022490
CAS Number: 473717-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR