tert-Butyl (2-(2-(prop-2-yn-1-yloxy)ethoxy)ethyl)carbamate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propargyl-PEG2-NHBoc
A761594-100MG
A761594-1G
A761594-250MG
A761594-5G
A761594-25G
76652-738EA
28.65
USD
76652-738
76652-740
76652-742
77599-034
77622-882
tert-Butyl (2-(2-(prop-2-yn-1-yloxy)ethoxy)ethyl)carbamate 95%
tert-Butyl (2-(2-(prop-2-yn-1-yloxy)ethoxy)ethyl)carbamate
Caution: for Research Use Only
Formula:
C₁₂H₂₁NO₄ MW: 243.2994 g/mol |
MDL Number:
MFCD22418469 CAS Number: 869310-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...