Titanium(IV) chloride 1 mol/l in toluene, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Titanium tetrachloride

37816-1000 378168000
TS37816-1000EA 63 USD
TS37816-1000 TS378168000
Titanium(IV) chloride 1 mol/l in toluene, AcroSeal™
Titanium (IV) chloride
Formula: Cl₄Ti
MW: 189.69
Boiling Pt: 136
Melting Pt: 136.2
Density: 0.964 g/cm³
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00011267
CAS Number: 7550-45-0
EINECS: 231-441-9
UN: 3286
ADR: 3 II
Merck Index: 13,09555

Order Now

Specification Test Results

Appearance Clear orange Solution
Titration Argentometric 0.95 to 1.1 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR