Hydrogen chloride 1 N in diethyl ether, AcroSeal™, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
36846-1000
368468000
TS36846-1000EA
64
USD
TS36846-1000
TS368468000
Hydrogen chloride 1 N in diethyl ether, AcroSeal™, pure
Hydrogen chloride
Formula:
HCl MW: 36.46 Density: 0.74 g/cm³ Flash Pt: –40 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 2924 ADR: 3 I |
Specification Test Results
Appearance | Clear colorless Solution |
Strength | 0.95 to 1.10 N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...