(-)-Terpinen-4-ol 97% enantiomers_sum
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(-)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (R)-1-Isopropyl-4-methylcyclohex-3-enol, (-)-4-Hydroxy-4-isopropyl-1-methyl-1-cyclohexene, (-)-p-Menth-1-en-4-ol
27043-0250
270431000
TS27043-0250EA
70
USD
TS27043-0250
TS270431000
(-)-Terpinen-4-ol 97% enantiomers_sum
(-)-Terpinen-4-ol
Appearance: Clear colorless to yellow Liquid
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 212 °C (760 mmHg) Density: 0.933 g/cm³ Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167108 CAS Number: 20126-76-5 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4760 to 1.4820 (20 °C, 589 nm) |
Density | (20/4°C) 0.930 to 0.936 g/ml |
GC | ≥96.0 % (sum of enantiomers) |
Specific optical rotation | -19° to -31° (20 °C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...