1-Hexene 97%

Supplier: Thermo Scientific Chemicals
Danger

21321-0010 213210025 213210250 213211000
TS21321-0010EA 51 USD
TS21321-0010 TS213210025 TS213210250 TS213211000
1-Hexene 97%
1-Hexene
Formula: C₆H₁₂
Boiling Pt: 62…65 °C (760 mmHg)
Melting Pt: –140 °C
Density: 0.678 g/cm³
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009505
CAS Number: 592-41-6
EINECS: 209-753-1
UN: 2370
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3867 to 1.3887 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR