Sulfur 99.5%, resublimed

Supplier: Thermo Scientific Chemicals
Warning

20125-0250 201250025
TS20125-0250EA 47 USD
TS20125-0250 TS201250025
Sulfur 99.5%, resublimed
Sulphur
Formula: S
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 114 °C
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059

Order Now

Specification Test Results

Appearance Yellow Powder
Assay ≥99.4 %
Sulfated ash ≤0.2 %
Free acid (H2SO4) ≤0.02 %
Free alkali (NaOH) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR