Germanium 99.999% (trace metals basis), powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
19311-0500
193110100
TS19311-0500EA
673
USD
TS19311-0500
TS193110100
Germanium 99.999% (trace metals basis), powder
Germanium
Formula:
Ge Boiling Pt: 2830 °C (1013 hPa) Melting Pt: 937 °C Density: 5.35 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 UN: 3089 ADR: 4.1,III Merck Index: 12,04415 |
Specification Test Results
Appearance | Grey to black Glistening crystalline powder |
Total trace metal impurities | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...