Poly(vinyl chloride), carboxylated, M.W. ∼220000 aver. carboxyl content 1.8 wt%
Supplier: Thermo Scientific Chemicals
18331-1000
183312500
TS18331-1000EA
104
USD
TS18331-1000
TS183312500
Poly(vinyl chloride), carboxylated, M.W. ∼220000 aver. carboxyl content 1.8 wt%
Poly(vinyl chloride), carboxylated
Formula:
(C₅H₇ClO₂)n Density: 1.39 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00084460 CAS Number: 25702-80-1 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | 5.0 to 11.0 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...