ADC® Adtemp Mini 432 Non-Contact Thermometer
Supplier: AMERICAN DIAGNOSTIC CORP CP
ADC®
432
470349-514CS 2958.99 USD
470349-514
ADC® Adtemp Mini 432 Non-Contact Thermometer
Thermometers Digital Thermometers
Non-contact thermometer.
  • Accurate readings In about 1 second
  • Compact, lightweight size
  • No skin contact required
  • Color-coded LCD display with fever warning
  • °F/°C Switchable display
  • Clips to retractable, breakaway lanyard (included) so it's always on hand
  • 2-year warranty
The Adtemp Mini 432 is compact, lightweight, and fast. Offering an ingenious design and incredible portability - along with the safety and convenience of a non-contact instrument - the 432 is sure to become an essential instrument for busy professionals.