ADC® Adscope 606
Supplier: AMERICAN DIAGNOSTIC CORP CP
606BK 606BD 606MCA 606MRS 606N 606PBCA 606RB
470349-492CS 583.99 USD
470349-492 470350-472 470350-386 470350-492 470350-422 470350-476 470350-394
ADC® Adscope 606
Stethoscopes
Ultra-lite cardiology stethoscope.
  • Inspected, assembled, and packaged in the USA
  • Lifetime warranty covers all parts
  • Scope ID tag included
Cardiology scope performance at a fraction of the weight, now with AFD technology. Combination chest piece precisely CNC machined to exacting tolerances from ultra-lightweight aluminum alloy with an extra-deep bell for unsurpassed low-frequency response. The ultra-sensitive diaphragm for greater amplification and crisper high-frequency transmission. AFD technology further enhances acoustic response. Use light pressure for low frequencies, firm pressure for high frequencies. Includes accessory kit with two spare pairs of Adsoft™ Plus ear tips and a Scope ID tag.