ADC® Adscope 615
Supplier: AMERICAN DIAGNOSTIC CORP CP
615BK 615PBCA 615R 615G 615LB 615LV 615MCA 615MCB 615MG 615MRS 615N 615O 615P 615BD 615DG 615FG 615FL 615FRB 615FS 615FV 615RB 615TL 615TQ 615V
470349-508CS 866.69 USD
470349-508 470350-486 470350-280 470350-564 470350-362 470350-326 470350-410 470350-222 470350-442 470350-372 470350-430 470350-470 470350-526 470350-494 470350-458 470350-550 470350-256 470350-444 470350-270 470350-390 470350-214 470350-274 470350-514 470350-290
ADC® Adscope 615
Stethoscopes
Platinum clinician stethoscope.
  • Inspected, assembled, and packaged in the USA
  • Lifetime warranty covers all parts
  • Scope ID tag included
With AFD technology and an oversized ovoid chest piece, the 615 is our finest clinician series stethoscope. ADC’s AFD technology provides the acoustic response of a traditional bell and diaphragm in a convenient one-sided chest piece. Light pressure for low, bell-type frequency, firm pressure for high, diaphragm-type frequency. The sculpted chest piece is machined from zinc alloy plated with an attractive satin stainless finish and includes a non-chill diaphragm retaining rim for patient comfort.