6-Fluorescein dT phosphoramidite

Supplier: AAT Bioquest

6200 6201
76644-680EA 130 USD
76644-680 76644-682
6-Fluorescein dT phosphoramidite

Fluorescein-dT phosphoramidite provides a valuable tool to add the fluorescein tag to a desired location of an oligo sequence. For efficiently labeling an oligonucleotide.


  • Spectral profile is compatible with all major fluorescence platforms


Fluorescein-dT phosphoramidite contains a DMT group to allow quantification of coupling. It can be inserted into a target sequence as a replacement for a dT residue.


It is complimentary to the commonly used 6-FAM phosphoramidite that contains no DMT group and can only be added at the 5'-terminus, thus terminating synthesis.


Packaging: Plastic vial.


Caution: For research use only.

Formula: C₇₉H₈₉N₆O₁₇P
CAS Number: 289712-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR