Reusable Ear Specula, Welch Allyn®
Supplier: Healthcare Products
WEL 22002 WEL 22003 WEL 22004 WEL 22005 WEL 22009
470347-296EA 11.65 USD
470347-296 470347-032 470347-104 470347-036 470347-300
Reusable Ear Specula, Welch Allyn®
Specula
Reusable specula are lightweight yet durable, and may be autoclaved, boiled, or sterilized in solution
Polypropylene base with soft, flexible tip provides improved seal for pneumatic otoscopy.