Acetonitrile

Supplier: MilliporeSigma

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

1.03725.1002 1.03725.2002
EM1.03725.1002EA 347 USD
EM1.03725.1002 EM1.03725.2002
Acetonitrile
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Flash Pt: 2
MDL Number: MFCD00001878
CAS Number: 75-05-8
Merck Index: 13,00071

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR