TI-Innovator Temperature and Humidity Module
Supplier: D&H
TI-Innovator
TEMPHUMIDMOD5PK
470348-850EA 39.99 USD
470348-850
TI-Innovator Temperature and Humidity Module
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Ambient temperature and humidity sensor for your TI-Innovator hub system.
  • Collect temperature and pressure data as inputs for programming
  • Real-world applications for your TI-Innovator hub
The TI-Innovator temperature & humidity module is a combined sensor that measures both the ambient temperature and relative humidity. It comes mounted on a printed circuit board for easy handling, with a cable to connect it to the TI-Innovator hub for programming. Build and program your Internet of Things (IoT) in your classroom using the TI-Innovator hub system and your existing TI-Nspire or TI-84 Plus CE technology.
Delivery information: Pack includes five temperature and humidity (DHT) sensors and cables.