BioClean™ Nitrile RABS/Isolator Sleeve/Glove System, Ansell
Supplier: Ansell Healthcare
GSG10NIT80 GSG10NITXLMA
76316-264CS 5697.71 USD
76316-264 76316-268
BioClean™ Nitrile RABS/Isolator Sleeve/Glove System, Ansell
Gloves
Validated sterile Nitrile RABS/Isolator Sleeve/Glove System for 203 mm (8") ports.
  • Tested against ASTM D6978-05 for handling chemo drugs
  • Ultra-clean surface ensures product protection
  • 100% visually inspected 
  • Fits 203 mm (8") port
  • Autoclavable compatible (sleeve only)
Clean and sterile sleeve/glove system features a nitrile sleeve attached to a PCP glove by a channel ring and O-ring. Sold by the pair, individually packaged.
Certifications: ASTM D6978, ISO 11137-1:2006, EN 420:2003 + A1:2009, EN 421:2010, EN ISO 374-1:2016, EN ISO 374-5:2016,EN 455 Part 2, Category III.
Packaging: Individually packed glove/sleeve and channel ring/O-ring assembly packed in inner PE bag; two inner bags (two systems) packed per outer PE bag; 10 outer bags (20 systems) per lined white corrugated carton.