AlphaTec® CSM Isolator Gloves, 85-301 Series, Ansell
Supplier: Ansell Healthcare
85301095 85301110
76408-464CS 5738.09 USD
76408-464 76408-466
AlphaTec® CSM Isolator Gloves, 85-301 Series, Ansell
Gloves
Non-clean/non sterile CSM RABS/Isolator gloves for use in 203 mm (8") ports.
  • Designed for high resistance against concentrated acids and bases
  • Port size: 203 mm (8")
  • Soft, flexible CSM material designed for ease of use
  • Resists ageing from exposure to oxygen, UV rays, and ozone
  • White colouring for easy detection of contamination
AlphaTec® CSM RABS and Isolator gloves are designed for high resistance against concentrated acids and bases. They are made from soft, flexible material designed for ease of use.
Certifications: CE 0493, EN ISO 374-1:2016,More Product Information