HYDRANAL™ - NEXTGEN Coulomat C-FA, Catholyte for Coulometric Karl Fischer Titration, Fluka™
Supplier: Honeywell Research Chemicals
34470-50ML
34470-6X50ML
BJ34470-50MLEA
381
USD
BJ34470-50ML
BJ34470-6X50ML
HYDRANAL™ - NEXTGEN Coulomat C-FA, Catholyte for Coulometric Karl Fischer Titration, Fluka™
Karl Fischer Titrants and Reagents
Catholyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, Acetonitrile-based, free of alcohols (free of CMR substances)
- Assay of water Max. 30 ppm
- Number of samples Min. 3
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)