Heat Sealing Foils, Pierceable
Supplier: AZENTA US, INC
4TI-05381/RA 4TI-0538-SLR 4TI-0539 4TI-0539/RA 4TI-0539/S 4TI-0538 4TI-0538/S 4TI-05381
76637-700CS 415.72 USD
76637-700 76637-710 76637-748 76637-874 76637-728 76637-934 76637-858 76637-770
Heat Sealing Foils, Pierceable
Seals Microplate Seals
The strong heat sealing foil is peelable from COC plates, pierceable, suitable for PCR, sample shipping and compound storage.
  • Compatible with polypropylene and COC plates
  • Recommended for PCR, sample shipping, low and room temperature compound storage with DMSO and other organic solvents
  • Suitable for long term storage
  • Gives a weld seal to polypropylene plates
  • Good solvent resistance
  • Temperature range: –20 to +120 °C
  • Non sterile
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective.
The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.
Ordering information: Available as sheets for use with manual and semiautomated sealers. Available in multiple roll formats compatible with specified automated heat sealers. Gamma treatment available upon request.