Anti-Flk-1 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP076-01 RP076-05 RP076
76635-090EA 138.83 USD
76635-090 76635-092 76635-094
Anti-Flk-1 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 180 kDa protein, known as Flk-1, KDR or vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a cell membrane receptor kinase. Flk-1 is a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.