Anti-Glycophorin A Mouse Monoclonal Antibody [clone: JC159]
Supplier: Diagnostic Biosystems
MOB066-01 MOB066-05 MOB066
76633-206EA 108.26 USD
76633-206 76633-208 76633-210
Anti-Glycophorin A Mouse Monoclonal Antibody [clone: JC159]
Antibodies
This antibody is specific to erythroid cells at various stages of differentiation from the erythroblast to the mature red cell.