Bacto™ TC Yeastolate, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Bacto TC Yeastolate is an animal origin-free hydrolysate from the water-soluble portion of autolyzed yeast. This supplement contains a mixture of peptides, amino acids, carbohydrates (simple and complex), nucleosides, and vitamins.
- Mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), as well as vitamins
- Meets additional requirements for reduced endotoxin, clarity, and suitability for the biopharmaceutical industry
- Bacterial, insect, and mammalian cell culture
- Supplementation of TC yeastolate to CHO cell culture can enhance growth and mAb production
Customers have used this supplement for CHO-based production of biotherapeutic monoclonal antibodies used in oncological and rheumatological therapeutics and recombinant proteins. Additionally, customers have used this supplement in combination with a soy supplement, such as Difco Phytone, ultra-filtered (UF), to provide an optimal titer performance in mammalian cell based processes by providing additional vitamins and carbohydrates.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)