Bacto™ Neopeptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
211680
211681
76628-796EA
8809.75
USD
76628-796
76628-820
Bacto™ Neopeptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology
Dehydrated Media for Microbiology
Bacto Neopeptone is an enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation.
- Enzymatic digest of bovine (milk) and porcine protein
- Animal origin nutritional supplement
- Contains a wide variety of peptide sizes in combination with vitamins, nucleotides, and minerals
- Recommended for use in media for detection of fungi
- Reported to provide nutrients for support of Spirochetes and Protozoa
- Microbial fermentation
- Production of vaccines for animal health applications
Customers have used Bacto Neopeptone as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A Streptococci, as well as Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health applications.