Phytone™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
292450
211906
76628-790EA
2644.04
USD
76628-790
76628-740
76628-776
Phytone™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology
Dehydrated Media for Microbiology
Phytone Peptone is an animal origin-free (AOF) enzymatic digest of soy. Due to the naturally high concentrations of vitamins and carbohydrates in soy, Phytone Peptone is a nutritious and excellent carbohydrate source.
- Enzymatic digests of soybean meal/flour
- Retains the high vitamin and high carbohydrate content of the soy plant tissue
- It is an excellent plant peptone for the cultivation of fungi and fastidious types of bacteria, such as members of the Clostridium and Neisseria genera
- It has been used in mammalian cell culture applications due to its high carbohydrate content
Customers have successfully used this supplement in CHO-based production of monoclonal antibodies for oncological and rheumatological therapeutics, as well as with many different microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines.
Certifications: LUO