Motic PX43MET Inverted Industrial Microscopes
Supplier: Motic
1100105400011 PA43-MT-BD-S6
76637-488EA 10284.75 USD
76637-488 76637-490
Motic PX43MET Inverted Industrial Microscopes
Microscopes
Motic’s new PX43MET is a professional inverted metallurgical microscope designed to inspect polished cross-sections as well as larger, bulkier materials samples. The microscope is a best fit for automotive, machinery, and metal fabrication industries and similar fields.
  • Trinocular head Siedentopf type 45° inclined, 360° swiveling (light split 100:0/20:80)
  • Widefield high eyepoint eyepiece 10×/20 mm (paired), focusable
  • Side facing quintuple nosepiece with DIC slot
  • Mechanical stage with metal stage insert
This full featured microscope is capable of (1) DIC for better depth imaging (2) polarization (3) brightfield and darkfield contrast methods (4) advanced measurement, image processing and documentation through a Moticam and the Motic snalysis software.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions.