Motic PSM1000 Upright Industrial Microscopes
Supplier: Motic
1100101700012 1100101700032 1100101700062 1100101700071
76637-470EA 7103.21 USD
76637-470 76637-472 76637-474 76637-476
Motic PSM1000 Upright Industrial Microscopes
Microscopes
The PSM-1000 microscope is an OEM-ready instrument for best possible imaging results of flat, reflective specimen. Metallurgical samples as well as semiconductors can be treated with plan apochromatic LWD objectives of stupendous resolution power.
  • PSM1000: trinocular head [optical split - 50:50 / 0:100]
  • PSM1000E: trinocular tilting head [optical split - 50:50 / 0:100]
  • Adjustable widefield high eyepoint 10×/F.N.24
  • 3-lens changeover turret [1XUV, 1XIR, 2XVIS]
A large field of view with an erect, true sided image allows fast orientation and manipulation work. Single centration of each objective position perfectly keeps the details of the viewing field. Optional objectives with parfocality adjustment capability additionally optimize ease of use when changing magnification. The high transmittance of the objectives and tube lens turret from NUV (355 nm) to NIR (1064 nm) facilitates the implementation of the PSM-1000 into laser probe stations. The smooth focus drive allows finest focusing even under high magnifications.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions.