2-Fluoro-5-methyl-4-nitrobenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC53482-1G PC53482-250MG
76624-316EA 454.32 USD
76624-316 76624-318
2-Fluoro-5-methyl-4-nitrobenzenesulfonyl chloride
2-Fluoro-5-methyl-4-nitrobenzenesulfonyl chloride
Formula: C₇H₅ClFNO₄S
Storage Temperature: Ambient
MDL Number: MFCD28797548
CAS Number: 1803585-04-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR