4-Bromo-5-chloro-2-fluorobenzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC53319-1G PC53319-5G
76624-306EA 242.05 USD
76624-306 76624-308 76624-310
4-Bromo-5-chloro-2-fluorobenzenesulfonyl chloride 97%
4-Bromo-5-chloro-2-fluorobenzenesulfonyl chloride
Formula: C₆H₂BrCl₂FO₂S
Storage Temperature: Ambient
MDL Number: MFCD12026216
CAS Number: 1208075-41-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR