2,4-Difluoro-3-(trifluoromethoxy)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC508033-1G PC508033-250MG PC508033-5G
76626-872EA 368.76 USD
76626-872 76626-874 77416-414
2,4-Difluoro-3-(trifluoromethoxy)benzonitrile
2,4-Difluoro-3-(trifluoromethoxy)benzonitrile
Formula: C₈H₂F₅NO
MW: 223.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28791003
CAS Number: 1806314-81-7
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR