2-Bromo-5-chloro-3-fluoropyridine

Supplier: APOLLO SCIENTIFIC
Warning

PC5924-1G PC5924-5G
76603-348EA 20.26 USD
76603-348 76603-350
2-Bromo-5-chloro-3-fluoropyridine
2-Bromo-5-chloro-3-fluoropyridine
Formula: C₅H₂BrClFN
Boiling Pt: 77…78 °C/7mmHg
Melting Pt: 28…30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044268
CAS Number: 514797-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR