Hexadecafluorosebaconitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Perfluorosebaconitrile

PC6213-1G PC6213-25G PC6213-5G
76627-264EA 62.14 USD
76627-264 76627-266 76627-268
Hexadecafluorosebaconitrile 97%
Hexadecafluorosebaconitrile
Formula: C₁₀F₁₆N₂
MW: 452.09 g/mol
Boiling Pt: 147…148 °C
Density: 1.673 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042366
CAS Number: 2342-09-8
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR