2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC8450-500MG PC8450-1G
76609-408EA 20.26 USD
76609-408 76609-406
2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone 97%
2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone
Formula: C₅H₅F₃N₄O
MW: 194.12 g/mol
Melting Pt: 217…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00068010
CAS Number: 95095-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR