3-Fluoro-4-nitrobenzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Bromomethyl)-2-fluoro-1-nitrobenzene

PC4404-1G PC4404-5G
76623-582EA 192.61 USD
76623-582 76623-584 76623-586
3-Fluoro-4-nitrobenzyl bromide 97%
3-Fluoro-4-nitrobenzyl bromide
Formula: C₇H₅BrFNO₂
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD03094237
CAS Number: 131858-37-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR