7-Hydroxy-4-(trifluoromethyl)coumarin 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Trifluoromethyl)umbelliferone, TFMU

PC4866-1G PC4866-5G
76618-396EA 22.42 USD
76618-396 76618-398
7-Hydroxy-4-(trifluoromethyl)coumarin 97%
7-Hydroxy-4-(trifluoromethyl)coumarin
Formula: C₁₀H₅F₃O₃
Melting Pt: 178…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00037578
CAS Number: 575-03-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR