4'-Bromo-2,2,2-trifluoroacetophenone 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC9923-100G PC9923-25G PC9923-5G
76610-676EA 239.4 USD
76610-676 76610-678 76610-680
4'-Bromo-2,2,2-trifluoroacetophenone 99%
4'-Bromo-2,2,2-trifluoroacetophenone
Formula: C₈H₄BrF₃O
Boiling Pt: 95 °C/4mmHg
Melting Pt: 26…30 °C
Density: 1.662 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00191862
CAS Number: 16184-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR