2,2,2-Trifluoroethyl trifluoromethanesulfonate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC3903-25G PC3903-5G PC3903-100G
76623-480EA 26.48 USD
76623-480 76623-482 76623-478
2,2,2-Trifluoroethyl trifluoromethanesulfonate 97%
2,2,2-Trifluoroethyl trifluoromethanesulphonate

Powerful trifluoroethylating agent; Useful for synthesis offluorinated amino acids, ask for references.

Formula: C₃H₂F₆O₃S
Boiling Pt: 91 °C
Density: 1.611 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00671579
CAS Number: 6226-25-1
EINECS: 458-390-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR