5-Amino-2-chloro-4-nitrobenzotrifluoride 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2-Nitro-4-chloro-5-(trifluoromethyl)aniline, 5-Amino-2-chloro-4-nitrobenzotrifluoride
PC5163-250MG
PC5163-5G
PC5163-1G
76600-108EA
20.87
USD
76600-108
76600-110
76600-106
5-Amino-2-chloro-4-nitrobenzotrifluoride 97%
5-Amino-2-chloro-4-nitrobenzotrifluoride
Formula:
C₇H₄ClF₃N₂O₂ MW: 240.5700 g/mol Melting Pt: 123…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094264 CAS Number: 167415-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...