2-Fluoro-5-nitrophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5009-5G PC5009-1G PC5009-25G
76604-918EA 33.62 USD
76604-918 76606-154 76606-292
2-Fluoro-5-nitrophenol 98%
2-Fluoro-5-nitrophenol
Formula: C₆H₄FNO₃
MW: 157.1000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09054718
CAS Number: 22510-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR