2-Bromo-1-ethylpyridinium tetrafluoroborate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: BEP, 2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate

PC8735-100G PC8735-25G PC8735-5G
76614-664EA 321.9 USD
76614-664 76614-666 76614-668
2-Bromo-1-ethylpyridinium tetrafluoroborate 98%
2-Bromo-1-ethylpyridinium tetrafluoroborate
Formula: C₇H₉BBrF₄N
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00060046
CAS Number: 878-23-9
EINECS: 212-900-2
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR