1H,1H,2H,2H-Tridecafluoro-1-octanol 97+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol

PC6147-25G PC6147-100G PC6147-250G
76603-508EA 20.99 USD
76603-508 76603-504 76603-506
1H,1H,2H,2H-Tridecafluoro-1-octanol 97+%
1H,1H,2H,2H-Tridecafluoro-1-octanol
Formula: C₈H₅F₁₃O
MW: 364.11 g/mol
Boiling Pt: 91…93 °C/36mmHg
Density: 1.678 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00042143
CAS Number: 647-42-7
EINECS: 211-477-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR